Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142523.1 | NP_001135995.1 | 10 | 1163 | ||
NM_007199.2 | NP_009130.2 | 11 | 1346 | |||
GenBank mRNA | AF113136.1 | - | 11 | 1307 | ||
AK298645.1 | - | 10 | 1104 | |||
AK314040.1 | - | 11 | 1328 | |||
BC057800.1 | - | 11 | 1321 | 30335802 | ||
BC069388.1 | - | 11 | 1294 | 7262303 |