Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142523.1 | NP_001135995.1 | 11 | 1314 | ||
NM_007199.2 | NP_009130.2 | 12 | 1497 | |||
GenBank mRNA | AF113136.1 | - | 12 | 1458 | ||
AK298645.1 | - | 11 | 1255 | |||
AK314040.1 | - | 12 | 1479 | |||
BC057800.1 | - | 12 | 1472 | 30335802 | ||
BC069388.1 | - | 12 | 1445 | 7262303 |