Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016248.3 | NP_057332.1 | 8 | 3044 | ||
XM_005266247.1 | XP_005266304.1 | 8 | 3045 | |||
XM_005266248.3 | XP_005266305.1 | 7 | 2938 | |||
XM_005266249.3 | XP_005266306.1 | 8 | 3045 | |||
XM_005266250.3 | XP_005266307.1 | 8 | 3045 | |||
XM_011534903.2 | XP_011533205.1 | 8 | 3041 | |||
XM_011534904.1 | XP_011533206.1 | 8 | 3237 | |||
XM_011534905.1 | XP_011533207.1 | 7 | 2995 | |||
XM_011534906.2 | XP_011533208.1 | 7 | 3213 | |||
XM_017020381.1 | XP_016875870.1 | 8 | 3095 | |||
XM_017020382.1 | XP_016875871.1 | 6 | 2885 | |||
XM_017020383.1 | XP_016875872.1 | 6 | 3073 | |||
GenBank mRNA | AB014529.2 | - | 1 | 984 | ||
AF176555.1 | - | 8 | 3044 |