Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007210.3 | NP_009141.2 | 8, 9 | 1678 | ||
XM_005268607.1 | XP_005268664.1 | 8, 9 | 1937 | |||
XM_006719214.2 | XP_006719277.1 | 7, 8 | 1512 | |||
XM_011537819.2 | XP_011536121.1 | 9, 10 | 1893 | |||
XM_011537822.2 | XP_011536124.1 | 8, 9 | 1614 | |||
XM_017018744.1 | XP_016874233.1 | 8, 9 | 1751 | |||
XM_017018745.1 | XP_016874234.1 | 9, 10 | 1736 | |||
GenBank mRNA | AB265820.1 | - | 7, 8 | 1563 | ||
AK025961.1 | - | 1, 2 | 87 | |||
AK092496.1 | - | 1, 2 | 1084 | |||
AK296852.1 | - | 9, 10 | 1584 | |||
BC035822.1 | - | 8, 9 | 1681 | 5552059 | ||
BC114505.1 | - | 6, 7 | 1357 | 40006926 | ||
Y08565.1 | - | 6, 7 | 1357 |