Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007210.3 | NP_009141.2 | 9 | 1774 | ||
XM_005268607.1 | XP_005268664.1 | 9 | 2033 | |||
XM_006719214.2 | XP_006719277.1 | 8 | 1608 | |||
XM_011537819.2 | XP_011536121.1 | 10 | 1989 | |||
XM_011537822.2 | XP_011536124.1 | 9 | 1710 | |||
XM_017018744.1 | XP_016874233.1 | 9 | 1847 | |||
XM_017018745.1 | XP_016874234.1 | 10 | 1832 | |||
GenBank mRNA | AB265820.1 | - | 8 | 1659 | ||
AK025961.1 | - | 2 | 183 | |||
AK074658.1 | - | 8 | 1672 | |||
AK092496.1 | - | 2 | 1180 | |||
AK296852.1 | - | 10 | 1680 | |||
BC035822.1 | - | 9 | 1777 | 5552059 | ||
BC114505.1 | - | 7 | 1453 | 40006926 | ||
Y08565.1 | - | 7 | 1453 |