Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164746.1 | NP_001158218.1 | 3 | 1093 | ||
NM_001164747.1 | NP_001158219.1 | 4 | 838 | |||
NM_001164748.1 | NP_001158220.1 | 3 | 772 | |||
NM_007211.4 | NP_009142.2 | 4 | 866 | |||
XM_006719027.2 | XP_006719090.1 | 3 | 640 | |||
XM_006719028.1 | XP_006719091.1 | 3 | 1044 | |||
XM_017018746.1 | XP_016874235.1 | 3 | 680 | |||
GenBank mRNA | AB093206.1 | - | 3 | 745 | ||
AK290055.1 | - | 4 | 838 | |||
AK292333.1 | - | 3 | 1087 | |||
AY665468.1 | - | 3 | 695 | |||
BC030021.1 | - | 4 | 868 | 5266519 | ||
U82396.1 | - | 2 | 666 |