Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007214.4 | NP_009145.1 | 10, 11 | 1229 | ||
XM_011535399.2 | XP_011533701.1 | 10, 11 | 978 | |||
XM_017010218.1 | XP_016865707.1 | 10, 11 | 1062 | |||
GenBank mRNA | AF100141.1 | - | 10, 11 | 960 | ||
AJ011779.1 | - | 10, 11 | 1058 | |||
AK001465.1 | - | 11, 12 | 1092 | |||
AK027374.1 | - | 7, 8 | 600 | |||
AK074835.1 | - | 10, 11 | 1045 | |||
AL137338.1 | - | 7, 8 | 576 | |||
BC023598.2 | - | 10, 11 | 1140 | |||
BC047221.1 | - | 10, 11 | 1098 | 5277083 | ||
BC048287.1 | - | 10, 11 | 1085 | 5212007 | ||
KU178551.1 | - | 10, 11 | 960 |