Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007214.4 | NP_009145.1 | 12 | 1348 | ||
XM_011535399.2 | XP_011533701.1 | 12 | 1097 | |||
XM_017010218.1 | XP_016865707.1 | 12 | 1177 | |||
GenBank mRNA | AF100141.1 | - | 12 | 1079 | ||
AJ011779.1 | - | 12 | 1177 | |||
AK001465.1 | - | 13 | 1211 | |||
AK027374.1 | - | 9 | 719 | |||
AK074835.1 | - | 12 | 1164 | |||
AL137338.1 | - | 9 | 695 | |||
BC023598.2 | - | 12 | 1259 | |||
BC047221.1 | - | 12 | 1217 | 5277083 | ||
BC048287.1 | - | 12 | 1204 | 5212007 | ||
KU178551.1 | - | 12 | 1079 |