Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007214.4 | NP_009145.1 | 8, 9 | 993 | ||
XM_011535399.2 | XP_011533701.1 | 8, 9 | 742 | |||
XM_017010218.1 | XP_016865707.1 | 8, 9 | 826 | |||
GenBank mRNA | AF100141.1 | - | 8, 9 | 724 | ||
AJ011779.1 | - | 8, 9 | 822 | |||
AK001465.1 | - | 9, 10 | 856 | |||
AK027374.1 | - | 5, 6 | 364 | |||
AK074835.1 | - | 8, 9 | 809 | |||
AL137338.1 | - | 5, 6 | 340 | |||
BC023598.2 | - | 8, 9 | 904 | |||
BC047221.1 | - | 8, 9 | 862 | 5277083 | ||
BC048287.1 | - | 8, 9 | 849 | 5212007 | ||
KU178551.1 | - | 8, 9 | 724 |