Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007216.3 | NP_009147.3 | 19 | 2998 | ||
NM_181507.1 | NP_852608.1 | 20 | 3155 | |||
NM_181508.1 | NP_852609.1 | 19 | 2811 | |||
XM_011519862.1 | XP_011518164.1 | 20 | 3146 | |||
XM_011519863.1 | XP_011518165.1 | 20 | 3054 | |||
XM_011519864.1 | XP_011518166.1 | 20 | 2986 | |||
XM_011519865.1 | XP_011518167.1 | 19 | 3368 | |||
XM_011519866.1 | XP_011518168.1 | 19 | 3019 | |||
XM_011519867.1 | XP_011518169.1 | 19 | 2885 | |||
XM_011519868.2 | XP_011518170.1 | 19 | 3126 | |||
XM_017017149.1 | XP_016872638.1 | 20 | 3054 | |||
XM_017017150.1 | XP_016872639.1 | 20 | 2986 | |||
XM_017017151.1 | XP_016872640.1 | 19 | 3204 | |||
XM_017017152.1 | XP_016872641.1 | 19 | 3368 | |||
XM_017017153.1 | XP_016872642.1 | 19 | 3204 | |||
XM_017017154.1 | XP_016872643.1 | 19 | 2885 | |||
GenBank mRNA | AB023234.1 | - | 19 | 2811 | ||
AF534400.1 | - | 19 | 2811 | |||
AF534401.1 | - | 20 | 3155 | |||
AF534402.1 | - | 19 | 2998 | |||
AJ131721.1 | - | 8 | 1329 | |||
AK024320.1 | - | 5 | 938 | |||
AK291663.1 | - | 20 | 3121 | |||
AK292436.1 | - | 19 | 2964 | |||
BC033640.1 | - | 19 | 2959 | 5502950 |