Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199653.1 | NP_001186582.1 | 3 | 341 | ||
NM_001199654.1 | NP_001186583.1 | 3 | 347 | |||
NM_007221.3 | NP_009152.2 | 3 | 341 | |||
GenBank mRNA | AF141310.1 | - | 3 | 304 | ||
AK124646.1 | - | 3 | 336 | |||
AK289490.1 | - | 3 | 322 | |||
AK290260.1 | - | 3 | 322 | |||
AK310694.1 | - | 2 | 216 | |||
AL080101.1 | - | 3 | 310 | |||
BC050735.1 | - | 3 | 296 | |||
BC056417.1 | - | 3 | 305 | |||
BC065031.1 | - | 3 | 320 | 6148250 | ||
DC310420.1 | - | 3 | 342 | |||
DT220098.1 | - | 3 | 329 |
Gene Symbol: PMF1-BGLAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199661.1 | NP_001186590.1 | 3 | 341 | ||
NM_001199662.1 | NP_001186591.1 | 3 | 341 | |||
NM_001199663.1 | NP_001186592.1 | 3 | 341 | |||
GenBank mRNA | BC033656.2 | - | 3 | 331 | ||
CD049192.1 | - | 3 | 349 | |||
CD388852.1 | - | 3 | 341 |