Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXPH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007224.3 | NP_009155.1 | 2 | 454 | ||
XM_017018747.1 | XP_016874236.1 | 2 | 392 | |||
GenBank mRNA | AK094231.1 | - | 3 | 659 | ||
AK290949.1 | - | 2 | 392 | |||
AY358614.1 | - | 2 | 445 | |||
BC036679.1 | - | 2 | 448 | 5264190 | ||
BC053581.1 | - | 2 | 378 | 5174266 |