Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FILIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042459.2 | NP_001035924.1 | 5 | 1658 | ||
NM_001282794.1 | NP_001269723.1 | 2 | 714 | |||
NM_014890.3 | NP_055705.2 | 2 | 714 | |||
NM_182909.3 | NP_878913.2 | 5 | 1658 | |||
XM_006713486.2 | XP_006713549.1 | 3 | 910 | |||
XM_011512371.2 | XP_011510673.1 | 3 | 776 | |||
GenBank mRNA | AF329092.1 | - | 5 | 1659 | ||
AF514867.1 | - | 4 | 1187 | |||
AF514868.1 | - | 4 | 1187 | |||
AF514869.1 | - | 4 | 1187 | |||
AK096195.1 | - | 3 | 509 | |||
AK225855.1 | - | 3 | 854 | |||
AX776292.1 | - | 5 | 1659 | |||
AX776294.1 | - | 4 | 1195 | |||
AX776296.1 | - | 4 | 1198 | |||
AX776298.1 | - | 4 | 1195 | |||
AY642382.1 | - | 4 | 1202 | |||
BC027860.1 | - | 2 | 645 | 5228947 | ||
EU531865.1 | - | 2 | 615 | |||
U53445.1 | - | 2 | 559 |