Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007240.2 | NP_009171.1 | 1, 2 | 384 | ||
GenBank mRNA | AF119226.1 | - | 1, 2 | 361 | ||
BC006286.1 | - | 1, 2 | 382 | 3958403 | ||
BT006633.1 | - | 1, 2 | 350 | |||
ES309317.1 | - | 1, 2 | 400 |