Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270707.1 | NP_001257636.1 | 1 | 246 | ||
NM_001270708.1 | NP_001257637.1 | 1 | 246 | |||
NM_001270709.1 | NP_001257638.1 | 1 | 246 | |||
NM_001270710.1 | NP_001257639.1 | 1 | 246 | |||
NM_007243.2 | NP_009174.1 | 2 | 443 | |||
NR_073065.1 | 1 | 246 | ||||
NR_073066.1 | 1 | 246 | ||||
GenBank mRNA | AF143676.1 | - | 1 | 89 | ||
AK075509.1 | - | 2 | 445 | |||
AK298152.1 | - | 1 | 103 | |||
AK315190.1 | - | 1 | 101 | |||
AY358411.1 | - | 1 | 110 | |||
BC021260.1 | - | 1 | 81 | |||
BC039865.1 | - | 1 | 109 | 6043663 | ||
BM847576.1 | - | 1 | 120 | |||
BM917173.1 | - | 1 | 103 | |||
BU159435.1 | - | 1 | 55 | |||
DA000315.1 | - | 1 | 101 | |||
JQ798186.1 | - | 1 | 109 | |||
JQ798187.1 | - | 1 | 109 |