Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014275.4 | NP_055090.1 | 2 | 587 | ||
NM_054013.3 | NP_463459.1 | 1 | 1027 | |||
XM_017008998.1 | XP_016864487.1 | 2 | 413 | |||
GenBank mRNA | AB000624.1 | - | 2 | 260 | ||
AK023137.1 | - | 1 | 1027 | |||
AK225490.1 | - | 1 | 61 | |||
AK225492.1 | - | 1 | 61 | |||
AL537522.3 | - | 2 | 587 | |||
AY358984.1 | - | 2 | 268 | |||
BC051835.1 | - | 2 | 463 | 6196058 |