Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007253.3 | NP_009184.1 | 11 | 1329 | ||
GenBank mRNA | AF133298.1 | - | 10 | 1272 | ||
AK092612.1 | - | 1 | 1376 | |||
AK096809.1 | - | 7 | 2983 | |||
AK300539.1 | - | 7 | 880 | |||
AK314685.1 | - | 11 | 1327 | |||
DB329701.1 | - | 3 | 482 |