Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007255.2 | NP_009186.1 | 3, 4 | 741 | ||
XM_006714816.3 | XP_006714879.1 | 3, 4 | 673 | |||
XM_017008999.1 | XP_016864488.1 | 3, 4 | 2864 | |||
GenBank mRNA | AB028600.1 | - | 3, 4 | 688 | ||
AF142675.1 | - | 3, 4 | 683 | |||
AJ005382.1 | - | 3, 4 | 648 | |||
AK022566.1 | - | 1, 2 | 1748 | |||
AK023506.1 | - | 3, 4 | 684 | |||
AK095882.1 | - | 3, 4 | 1345 | |||
AY358578.1 | - | 3, 4 | 724 | |||
BC007317.2 | - | 3, 4 | 741 | 3049998 | ||
BC062983.1 | - | 3, 4 | 708 | 4577572 | ||
BC072403.1 | - | 3, 4 | 730 | 6094074 |