Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007261.3 | NP_009192.2 | 2 | 645 | ||
XM_005256990.2 | XP_005257047.1 | 2 | 666 | |||
XM_005256991.4 | XP_005257048.1 | 2 | 668 | |||
XM_005256992.4 | XP_005257049.1 | 2 | 574 | |||
XM_006721656.3 | XP_006721719.1 | 2 | 682 | |||
GenBank mRNA | AF020314.1 | - | 2 | 401 | ||
AJ010101.1 | - | 2 | 353 | |||
AK309631.1 | - | 3 | 710 | |||
AK309636.1 | - | 2 | 352 | |||
BC032352.1 | - | 2 | 610 | 5218501 | ||
BM923070.1 | - | 656 |