Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256841.1 | NP_001243770.1 | 2 | 399 | ||
NM_007261.3 | NP_009192.2 | 3 | 738 | |||
XM_005256990.2 | XP_005257047.1 | 3 | 759 | |||
XM_005256991.4 | XP_005257048.1 | 3 | 761 | |||
XM_005256992.4 | XP_005257049.1 | 3 | 667 | |||
XM_006721656.3 | XP_006721719.1 | 3 | 775 | |||
XM_011524254.2 | XP_011522556.1 | 2 | 462 | |||
GenBank mRNA | AF020314.1 | - | 3 | 494 | ||
AF161346.1 | - | 2 | 125 | |||
AJ010101.1 | - | 3 | 446 | |||
AJ010102.1 | - | 2 | 107 | |||
AJ224864.1 | - | 3 | 719 | |||
AJ238323.1 | - | 3 | 446 | |||
AK309631.1 | - | 4 | 803 | |||
AK309636.1 | - | 3 | 445 | |||
BC032352.1 | - | 3 | 703 | 5218501 |