Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123377.1 | NP_001116849.1 | 7 | 649 | ||
NM_007262.4 | NP_009193.2 | 7 | 707 | |||
XM_005263424.3 | XP_005263481.1 | 7 | 727 | |||
GenBank mRNA | AB073864.1 | - | 6 | 544 | ||
AF021819.1 | - | 7 | 615 | |||
AK091679.1 | - | 1 | 2070 | |||
AK130886.1 | - | 6 | 542 | |||
AK312000.1 | - | 7 | 602 | |||
BC008188.2 | - | 7 | 628 | 2901102 | ||
D61380.2 | - | 6 | 550 | |||
EU794641.1 | - | 6 | 550 |