Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100431.1 | NP_001093901.1 | 2 | 401 | ||
NM_001184830.1 | NP_001171759.1 | 2 | 401 | |||
NM_001184831.1 | NP_001171760.1 | 2 | 401 | |||
NM_001257403.1 | NP_001244332.1 | 2 | 401 | |||
NM_007268.2 | NP_009199.1 | 2 | 401 | |||
XM_017029251.1 | XP_016884740.1 | 2 | 360 | |||
GenBank mRNA | AJ132502.1 | - | 2 | 319 | ||
AK075153.1 | - | 2 | 356 | |||
AK075171.1 | - | 2 | 383 | |||
AK293215.1 | - | 2 | 295 | |||
AK297050.1 | - | 2 | 356 | |||
AK298389.1 | - | 2 | 354 | |||
AK301417.1 | - | 2 | 356 | |||
AL542949.3 | - | 1 | 53 | |||
AL552873.3 | - | 2 | 345 | |||
AL553835.3 | - | 2 | 378 | |||
AY358341.1 | - | 2 | 392 | |||
BC010525.1 | - | 2 | 277 | 4151604 | ||
BP292528.1 | - | 2 | 398 |