Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144831.1 | NP_001138303.1 | 7 | 975 | ||
NM_001267700.1 | NP_001254629.1 | 6 | 861 | |||
GenBank mRNA | AF126021.1 | - | 7 | 950 | ||
AF150962.1 | - | 7 | 888 | |||
AF178980.1 | - | 7 | 864 | |||
AF279895.1 | - | 2 | 76 | |||
AK298217.1 | - | 6 | 781 | |||
AK301316.1 | - | 6 | 1051 | |||
BC013228.1 | - | 7 | 858 | |||
BC014766.1 | - | 7 | 845 | 3452360 | ||
BC110322.1 | - | 7 | 975 | 5540527 | ||
U72511.1 | - | 5 | 503 |