Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2AF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012478.1 | NP_001012496.1 | 4 | 1374 | ||
NM_007279.2 | NP_009210.1 | 4 | 1374 | |||
XM_006722994.2 | XP_006723057.1 | 5 | 1412 | |||
XM_011526410.1 | XP_011524712.1 | 5 | 1412 | |||
GenBank mRNA | AB451261.1 | - | 4 | 319 | ||
AB451387.1 | - | 4 | 319 | |||
BC008740.2 | - | 4 | 378 | 3346087 | ||
BC030574.2 | - | 4 | 1278 | 3687471 | ||
X64044.1 | - | 4 | 359 |