Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181503.2 | NP_852480.1 | 5, 6 | 495 | ||
XM_006719763.1 | XP_006719826.1 | 4, 5 | 626 | |||
GenBank mRNA | AF025438.1 | - | 4, 5 | 224 | ||
AF279901.1 | - | 6, 7 | 286 | |||
AK096810.1 | - | 3, 4 | 2135 | |||
BC020773.1 | - | 5, 6 | 256 | 4808747 | ||
BM697343.1 | - | 5, 6 | 495 |