Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007281.2 | NP_009212.1 | 2 | 497 | ||
XM_011531578.1 | XP_011529880.1 | 3 | 216 | |||
XM_017007678.1 | XP_016863167.1 | 3 | 210 | |||
GenBank mRNA | AJ224677.1 | - | 2 | 497 | ||
AK312146.1 | - | 2 | 146 | |||
AY359040.1 | - | 2 | 244 | |||
BC017583.2 | - | 2 | 276 | |||
CR542142.1 | - | 1 | 84 |