Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004439.1 | NP_001004439.1 | 12, 13 | 1513 | ||
XM_005254228.3 | XP_005254285.1 | 10, 11 | 1735 | |||
XM_011521363.2 | XP_011519665.1 | 10, 11 | 1465 | |||
GenBank mRNA | AF109681.1 | - | 12, 13 | 1498 | ||
AF111799.1 | - | 4, 5 | 478 | |||
AF137378.2 | - | 12, 13 | 1516 | |||
AK292445.1 | - | 12, 13 | 1522 | |||
AK304778.1 | - | 12, 13 | 1523 | |||
BX496541.1 | - | 4, 5 | 502 | |||
CN358985.1 | - | 2, 3 | 293 |