Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317960.1 | NP_001304889.1 | 11 | 1270 | ||
NM_012255.4 | NP_036387.2 | 11 | 1110 | |||
XM_017027722.1 | XP_016883211.1 | 11 | 1265 | |||
XM_017027723.1 | XP_016883212.1 | 11 | 1039 | |||
XM_017027724.1 | XP_016883213.1 | 11 | 1265 | |||
GenBank mRNA | AF064257.1 | - | 11 | 1063 | ||
AF152169.1 | - | 11 | 1013 | |||
AK001573.1 | - | 11 | 1080 | |||
AK302846.1 | - | 11 | 974 | |||
AK303312.1 | - | 10 | 921 | |||
AK314602.1 | - | 11 | 1049 | |||
AL136841.1 | - | 11 | 1036 | |||
AY382900.1 | - | 10 | 925 |