Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145365.1 | NP_001138837.1 | 2 | 1086 | ||
NM_014897.2 | NP_055712.1 | 2 | 941 | |||
GenBank mRNA | AB023141.1 | - | 2 | 941 | ||
AK292667.1 | - | 2 | 1076 | |||
BC139779.1 | - | 1 | 796 | 40122232 | ||
BC152440.1 | - | 2 | 941 | 8862563 | ||
CR933680.1 | - | 2 | 937 |