Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014907.2 | NP_055722.2 | 16 | 2438 | ||
XM_011517803.2 | XP_011516105.1 | 17 | 2840 | |||
XM_011517804.2 | XP_011516106.1 | 16 | 2759 | |||
XM_011517805.2 | XP_011516107.1 | 13 | 2211 | |||
XM_011517806.2 | XP_011516108.1 | 12 | 3447 | |||
XM_017014480.1 | XP_016869969.1 | 16 | 3119 | |||
XM_017014481.1 | XP_016869970.1 | 12 | 2814 | |||
XM_017014482.1 | XP_016869971.1 | 13 | 2812 | |||
GenBank mRNA | AB023184.1 | - | 16 | 2438 | ||
BC114965.1 | - | 16 | 2417 | 40082639 | ||
BX648967.1 | - | 16 | 2988 |