Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256053.1 | NP_001242982.1 | 25 | 3905 | ||
NM_014915.2 | NP_055730.2 | 25 | 3908 | |||
XM_006717423.2 | XP_006717486.1 | 26 | 4994 | |||
XM_006717425.3 | XP_006717488.1 | 26 | 4994 | |||
XM_011519416.2 | XP_011517718.1 | 26 | 4994 | |||
XM_017015928.1 | XP_016871417.1 | 26 | 4994 | |||
XM_017015929.1 | XP_016871418.1 | 27 | 4982 | |||
XM_017015930.1 | XP_016871419.1 | 26 | 4994 | |||
XM_017015931.1 | XP_016871420.1 | 26 | 4994 | |||
XM_017015932.1 | XP_016871421.1 | 26 | 4994 | |||
XM_017015933.1 | XP_016871422.1 | 26 | 4994 | |||
GenBank mRNA | AB028997.1 | - | 25 | 3883 | ||
AK128577.1 | - | 10 | 2468 | |||
BC130453.1 | - | 25 | 3849 | 40146484 |