Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113226.2 | NP_001106697.1 | 2 | 1313 | ||
NM_001113228.2 | NP_001106699.1 | 2 | 1313 | |||
NM_001312688.1 | NP_001299617.1 | 3 | 1595 | |||
NM_014917.3 | NP_055732.2 | 3 | 1595 | |||
XM_006710455.2 | XP_006710518.1 | 3 | 2229 | |||
XM_006710456.3 | XP_006710519.1 | 3 | 2229 | |||
XM_011541021.1 | XP_011539323.1 | 3 | 2229 | |||
XM_011541024.2 | XP_011539326.1 | 3 | 2229 | |||
XM_011541025.1 | XP_011539327.1 | 3 | 2229 | |||
XM_017000680.1 | XP_016856169.1 | 3 | 1595 | |||
XM_017000681.1 | XP_016856170.1 | 3 | 2548 | |||
XM_017000682.1 | XP_016856171.1 | 3 | 2229 | |||
XM_017000683.1 | XP_016856172.1 | 3 | 1595 | |||
XM_017000684.1 | XP_016856173.1 | 3 | 2548 | |||
XM_017000685.1 | XP_016856174.1 | 3 | 2548 | |||
XM_017000686.1 | XP_016856175.1 | 3 | 1496 | |||
GenBank mRNA | AB023193.1 | - | 3 | 1527 | ||
AK296533.1 | - | 3 | 1496 | |||
AK297659.1 | - | 2 | 602 | |||
AY358365.1 | - | 2 | 1053 | |||
BC030220.1 | - | 3 | 1390 | 5206594 | ||
BX538348.1 | - | 3 | 1506 | |||
KJ534901.1 | - | 2 | 788 |