Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079673.1 | NP_001073141.1 | 19 | 2437 | ||
NM_001278438.1 | NP_001265367.1 | 19 | 2480 | |||
NM_014923.4 | NP_055738.3 | 17 | 2061 | |||
NR_103528.1 | 19 | 2469 | ||||
XM_011534997.2 | XP_011533299.1 | 18 | 2151 | |||
XM_017020440.1 | XP_016875929.1 | 19 | 2322 | |||
XM_017020441.1 | XP_016875930.1 | 21 | 2943 | |||
XM_017020442.1 | XP_016875931.1 | 20 | 2475 | |||
XM_017020443.1 | XP_016875932.1 | 19 | 2795 | |||
GenBank mRNA | AB023187.2 | - | 17 | 1991 | ||
AJ749706.1 | - | 19 | 2439 | |||
AJ749707.1 | - | 17 | 2052 | |||
AK302415.1 | - | 17 | 2061 | |||
BC060816.1 | - | 10 | 1099 | |||
BC070072.1 | - | 10 | 1099 | |||
BC132812.1 | - | 19 | 2204 | 40146834 | ||
BC136617.1 | - | 19 | 2205 | 9020607 | ||
BC144301.1 | - | 19 | 2205 | 9052821 | ||
BX640739.1 | - | 19 | 2340 | |||
BX648141.1 | - | 19 | 2427 | |||
BX648706.1 | - | 21 | 2285 |