Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079673.1 | NP_001073141.1 | 26 | 3623 | ||
NM_001278438.1 | NP_001265367.1 | 26 | 3666 | |||
NM_014923.4 | NP_055738.3 | 24 | 3247 | |||
NR_103528.1 | 26 | 3655 | ||||
XM_011534997.2 | XP_011533299.1 | 25 | 3337 | |||
XM_017020440.1 | XP_016875929.1 | 26 | 3508 | |||
XM_017020441.1 | XP_016875930.1 | 28 | 4129 | |||
XM_017020442.1 | XP_016875931.1 | 27 | 3661 | |||
GenBank mRNA | AB023187.2 | - | 24 | 3177 | ||
AJ749706.1 | - | 26 | 3625 | |||
AJ749707.1 | - | 24 | 3238 | |||
AK172814.1 | - | 4 | 554 | |||
AK302415.1 | - | 24 | 3247 | |||
BC060816.1 | - | 17 | 2285 | |||
BC070072.1 | - | 17 | 2285 | |||
BC132812.1 | - | 26 | 3390 | 40146834 | ||
BC136617.1 | - | 26 | 3391 | 9020607 | ||
BC144301.1 | - | 26 | 3391 | 9052821 | ||
BX640739.1 | - | 26 | 3526 | |||
BX648141.1 | - | 26 | 3613 | |||
BX648706.1 | - | 28 | 3471 |