Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014924.4 | NP_055739.2 | 6 | 806 | ||
XM_011536563.2 | XP_011534865.1 | 4 | 510 | |||
GenBank mRNA | AK131251.1 | - | 6 | 806 | ||
AK292810.1 | - | 6 | 778 | |||
BC109090.1 | - | 6 | 791 | 40034574 |