Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEKHA6 SIRNA Assays ›
Gene Symbol:
PLEKHA6
Gene Name:
pleckstrin homology domain containing A6
Gene Aliases:
PEPP-3, PEPP3
Chromosome Location:
Chr.1: 204218851 - 204378376 on Build GRCh38
UniGene ID:
Hs.253146
Species:
Human
siRNA ID
HSS117794
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEKHA6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014935.4
NP_055750.2
9
1804
XM_005244966.4
XP_005245023.2
9
1671
XM_005244967.4
XP_005245024.2
9
1730
XM_005244968.4
XP_005245025.2
9
1670
XM_006711212.3
XP_006711275.1
9
1957
XM_006711213.3
XP_006711276.2
9
1731
XM_006711214.3
XP_006711277.2
9
1731
XM_006711215.3
XP_006711278.2
9
1731
XM_006711217.3
XP_006711280.2
9
1731
XM_006711219.3
XP_006711282.2
9
1731
XM_006711221.2
XP_006711284.1
9
2055
XM_006711222.3
XP_006711285.1
9
2040
XM_011509295.2
XP_011507597.1
9
1731
XM_011509296.2
XP_011507598.1
9
1731
XM_011509297.2
XP_011507599.1
11
2557
XM_011509298.2
XP_011507600.1
9
1840
XM_011509299.2
XP_011507601.2
9
1846
XM_011509301.2
XP_011507603.1
9
5736
XM_011509304.2
XP_011507606.1
9
1685
XM_017000687.1
XP_016856176.1
9
1731
XM_017000688.1
XP_016856177.1
9
1731
XM_017000689.1
XP_016856178.1
9
1671
XM_017000690.1
XP_016856179.1
9
5676
XM_017000691.1
XP_016856180.1
4
4129
XM_017000692.1
XP_016856181.1
4
4069
GenBank mRNA
AB023186.1
-
9
1791
BC152475.1
-
9
1791
8862612
DQ925685.1
-
1
77
DQ925686.1
-
1
77
DQ925687.1
-
1
77
DQ925688.1
-
1
77
DQ925689.1
-
1
77
DQ925690.1
-
1
77
DQ925691.1
-
1
77
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-82rcf:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline