Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014936.4 | NP_055751.1 | 2 | 964 | ||
GenBank mRNA | AB020686.1 | - | 1 | 753 | ||
AK291276.1 | - | 2 | 809 | |||
BC018054.1 | - | 2 | 934 | 4797659 |