Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303256.2 | NP_001290185.1 | 19 | 2332 | ||
NM_001303257.2 | NP_001290186.1 | 19 | 2332 | |||
NM_014941.3 | NP_055756.1 | 20 | 3198 | |||
XM_011530004.1 | XP_011528306.1 | 19 | 2557 | |||
XM_017028667.1 | XP_016884156.1 | 19 | 2557 | |||
XM_017028668.1 | XP_016884157.1 | 20 | 2652 | |||
GenBank mRNA | AB020659.1 | - | 20 | 3112 | ||
BC019257.1 | - | 18 | 1859 | 3503651 | ||
BC136782.1 | - | 20 | 2057 | 9020772 | ||
CR456469.1 | - | 18 | 1858 |