Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014023.3 | NP_054742.2 | 2 | 288 | ||
GenBank mRNA | AB023199.1 | - | 2 | 259 | ||
AK292591.1 | - | 2 | 251 | |||
AK307259.1 | - | 2 | 278 | |||
AL136827.1 | - | 2 | 266 | |||
BC018044.1 | - | 2 | 317 | 4794184 |