Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014023.3 | NP_054742.2 | 4 | 482 | ||
GenBank mRNA | AB023199.1 | - | 4 | 453 | ||
AK128194.1 | - | 3 | 3291 | |||
AK292591.1 | - | 4 | 445 | |||
AK307259.1 | - | 3 | 375 | |||
AL136827.1 | - | 4 | 460 | |||
BC018044.1 | - | 4 | 511 | 4794184 |