Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301015.1 | NP_001287944.1 | 18, 19 | 2346 | ||
NM_001301018.1 | NP_001287947.1 | 17, 18 | 1721 | |||
NM_001301027.1 | NP_001287956.1 | 15, 16 | 2092 | |||
NM_001301028.1 | NP_001287957.1 | 14, 15 | 2013 | |||
NM_014945.3 | NP_055760.1 | 19, 20 | 1992 | |||
XM_005268392.1 | XP_005268449.1 | 18, 19 | 1874 | |||
XM_005268393.1 | XP_005268450.1 | 17, 18 | 1826 | |||
XM_005268394.1 | XP_005268451.1 | 17, 18 | 1787 | |||
XM_005268395.1 | XP_005268452.1 | 16, 17 | 1688 | |||
XM_005268396.1 | XP_005268453.1 | 15, 16 | 1640 | |||
XM_017009238.1 | XP_016864727.1 | 15, 16 | 1782 | |||
GenBank mRNA | AB020650.1 | - | 19, 20 | 1905 | ||
AB075881.1 | - | 4, 5 | 1915 | |||
AK225769.1 | - | 18, 19 | 2214 | |||
AK289736.1 | - | 18, 19 | 1759 | |||
AL833021.1 | - | 12, 13 | 1100 | |||
BC001665.2 | - | 14, 15 | 1675 | 2988043 | ||
DQ413174.1 | - | 17, 18 | 1683 |