Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198850.1 | NP_001185779.1 | 11 | 1755 | ||
NM_001198851.1 | NP_001185780.1 | 13 | 1798 | |||
NM_001198852.1 | NP_001185781.1 | 10 | 1558 | |||
NM_014947.4 | NP_055762.3 | 11 | 1677 | |||
XM_005270632.4 | XP_005270689.1 | 10 | 1565 | |||
XM_006710458.3 | XP_006710521.1 | 11 | 1784 | |||
XM_006710459.3 | XP_006710522.1 | 9 | 1464 | |||
XM_011541026.2 | XP_011539328.1 | 12 | 1729 | |||
XM_017000693.1 | XP_016856182.1 | 10 | 1702 | |||
XM_017000694.1 | XP_016856183.1 | 9 | 1441 | |||
XM_017000695.1 | XP_016856184.1 | 10 | 1327 | |||
GenBank mRNA | AB028964.1 | - | 13 | 1798 | ||
AK027075.1 | - | 11 | 1755 | |||
AL157422.1 | - | 1 | 914 | |||
BC151828.1 | - | 11 | 1646 | 40148791 | ||
BC152441.1 | - | 13 | 1798 | 8862565 |