Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014949.3 | NP_055764.2 | 3 | 376 | ||
GenBank mRNA | AB020714.1 | - | 3 | 333 | ||
AK225714.1 | - | 3 | 347 | |||
AK289902.1 | - | 3 | 319 | |||
AY112680.1 | - | 3 | 307 | |||
AY112681.1 | - | 3 | 307 | |||
AY112682.1 | - | 3 | 307 | |||
BC027182.1 | - | 3 | 332 | |||
BC062637.1 | - | 3 | 331 | 5497581 | ||
BI560023.1 | - | 3 | 386 | |||
KU178555.1 | - | 3 | 307 | |||
KU178556.1 | - | 3 | 307 |