Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014957.2 | NP_055772.2 | 19 | 3248 | ||
XM_005250838.4 | XP_005250895.2 | 19 | 3139 | |||
XM_005250839.2 | XP_005250896.2 | 19 | 3139 | |||
XM_005250840.4 | XP_005250897.2 | 18 | 2983 | |||
XM_005250841.3 | XP_005250898.2 | 18 | 2983 | |||
XM_005250842.4 | XP_005250899.1 | 17 | 3000 | |||
XM_011516933.1 | XP_011515235.1 | 19 | 3139 | |||
XM_011516934.2 | XP_011515236.1 | 19 | 3139 | |||
XM_011516938.2 | XP_011515240.1 | 12 | 2294 | |||
XM_011516939.2 | XP_011515241.1 | 10 | 1817 | |||
XM_011516940.2 | XP_011515242.1 | 10 | 1794 | |||
XM_017013241.1 | XP_016868730.1 | 19 | 3141 | |||
XM_017013242.1 | XP_016868731.1 | 20 | 3502 | |||
XM_017013243.1 | XP_016868732.1 | 11 | 2254 | |||
GenBank mRNA | AB020677.2 | - | 14 | 2435 | ||
AK131505.1 | - | 2 | 917 | |||
BC133023.1 | - | 17 | 2835 | 40147045 | ||
BC144226.1 | - | 17 | 2835 | 9052746 |