Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267727.1 | NP_001254656.1 | 12 | 2163 | ||
NM_014960.4 | NP_055775.2 | 12 | 2334 | |||
XM_005257170.3 | XP_005257227.1 | 12 | 2347 | |||
XM_006721777.3 | XP_006721840.2 | 13 | 2442 | |||
XM_011524535.2 | XP_011522837.1 | 13 | 2440 | |||
XM_011524536.2 | XP_011522838.1 | 13 | 2270 | |||
XM_011524537.1 | XP_011522839.1 | 13 | 2191 | |||
XM_011524538.2 | XP_011522840.1 | 13 | 2442 | |||
XM_011524540.2 | XP_011522842.1 | 13 | 2462 | |||
XM_011524545.2 | XP_011522847.1 | 11 | 2264 | |||
XM_011524546.2 | XP_011522848.1 | 8 | 1113 | |||
XM_017024360.1 | XP_016879849.1 | 13 | 2280 | |||
XM_017024361.1 | XP_016879850.1 | 12 | 2347 | |||
XM_017024362.1 | XP_016879851.1 | 12 | 2286 | |||
XM_017024363.1 | XP_016879852.1 | 12 | 2192 | |||
XM_017024364.1 | XP_016879853.1 | 12 | 2188 | |||
XM_017024365.1 | XP_016879854.1 | 12 | 2176 | |||
XM_017024368.1 | XP_016879857.1 | 11 | 2068 | |||
GenBank mRNA | AB023218.1 | - | 11 | 1996 | ||
AK311327.1 | - | 3 | 737 | |||
AY358380.1 | - | 12 | 2334 | |||
BC012375.1 | - | 11 | 1577 | 3882163 |