Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIFAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204514.1 | NP_001191443.1 | 15 | 1966 | ||
NM_001204516.1 | NP_001191445.1 | 13 | 1670 | |||
NM_001204517.1 | NP_001191446.1 | 14 | 2911 | |||
NM_014970.3 | NP_055785.2 | 14 | 1802 | |||
XM_005244970.1 | XP_005245027.1 | 14 | 1805 | |||
XM_011509307.2 | XP_011507609.2 | 14 | 1805 | |||
GenBank mRNA | AK026471.1 | - | 14 | 1581 | ||
AK122897.1 | - | 14 | 2913 | |||
AK297368.1 | - | 13 | 1597 | |||
AK301912.1 | - | 7 | 769 | |||
AK303052.1 | - | 15 | 1923 | |||
AK315586.1 | - | 14 | 1716 | |||
BC028679.2 | - | 14 | 1759 | 4829354 | ||
BC048012.1 | - | 13 | 1714 | |||
U59919.1 | - | 14 | 1719 |