Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIFAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204514.1 | NP_001191443.1 | 21 | 2785 | ||
NM_001204516.1 | NP_001191445.1 | 19 | 2489 | |||
NM_001204517.1 | NP_001191446.1 | 20 | 3730 | |||
NM_014970.3 | NP_055785.2 | 20 | 2621 | |||
XM_005244970.1 | XP_005245027.1 | 21 | 2661 | |||
XM_011509307.2 | XP_011507609.2 | 21 | 2693 | |||
GenBank mRNA | AI085654.1 | - | 1 | 371 | ||
AK026471.1 | - | 20 | 2400 | |||
AK122897.1 | - | 20 | 3732 | |||
AK297368.1 | - | 19 | 2416 | |||
AK301912.1 | - | 13 | 1588 | |||
AK303052.1 | - | 21 | 2742 | |||
AK315586.1 | - | 20 | 2535 | |||
BC028679.2 | - | 20 | 2578 | 4829354 | ||
BC048012.1 | - | 19 | 2533 | |||
U59919.1 | - | 20 | 2538 |