Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306153.1 | NP_001293082.1 | 5 | 579 | ||
NM_001306154.1 | NP_001293083.1 | 4 | 814 | |||
NM_001306155.1 | NP_001293084.1 | 4 | 646 | |||
NM_001306156.1 | NP_001293085.1 | 5 | 811 | |||
NM_012073.4 | NP_036205.1 | 5 | 979 | |||
GenBank mRNA | AK289353.1 | - | 5 | 577 | ||
AK290393.1 | - | 5 | 620 | |||
AK293341.1 | - | 5 | 648 | |||
AK293437.1 | - | 5 | 520 | |||
AK301760.1 | - | 5 | 571 | |||
AK301985.1 | - | 6 | 537 | |||
AK302368.1 | - | 4 | 519 | |||
AK302383.1 | - | 4 | 644 | |||
AK303086.1 | - | 6 | 506 | |||
AK303147.1 | - | 2 | 157 | |||
AK316376.1 | - | 5 | 566 | |||
BC002971.1 | - | 5 | 554 | |||
BC006543.2 | - | 5 | 594 | 2824199 | ||
BC009454.1 | - | 2 | 98 | |||
BC035499.2 | - | 5 | 641 | 4329194 | ||
BC040027.1 | - | 5 | 611 | |||
D43950.2 | - | 5 | 596 | |||
FJ224303.1 | - | 5 | 730 |