Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007349.3 | NP_031375.3 | 19 | 3147 | ||
XM_005249539.1 | XP_005249596.1 | 20 | 3286 | |||
XM_011515982.2 | XP_011514284.1 | 19 | 3261 | |||
XM_011515983.1 | XP_011514285.1 | 19 | 3147 | |||
XM_017011881.1 | XP_016867370.1 | 20 | 4459 | |||
GenBank mRNA | AK093584.1 | - | 13 | 1840 | ||
AK123600.1 | - | 20 | 3286 | |||
AK126674.1 | - | 11 | 3226 | |||
AK293744.1 | - | 17 | 2979 | |||
BC008328.1 | - | 10 | 1071 | |||
BC033781.1 | - | 14 | 2388 | 4509264 | ||
BX538201.1 | - | 13 | 1923 | |||
BX640616.1 | - | 19 | 5045 | |||
U80735.1 | - | 15 | 2130 |