Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR7A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012067.2 | NP_036199.2 | 2 | 403 | ||
XM_011541046.2 | XP_011539348.1 | 2 | 979 | |||
XM_017000714.1 | XP_016856203.1 | 2 | 137 | |||
GenBank mRNA | AF040639.1 | - | 2 | 410 | ||
AJ271799.1 | - | 2 | 333 | |||
BC025709.1 | - | 2 | 389 | 5224626 | ||
BC031562.1 | - | 2 | 345 | 5184829 | ||
BC042420.1 | - | 2 | 369 | 5186889 | ||
BM920315.1 | - | 2 | 367 |